Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 488)

Lieferant: Agrisera
AS101519
AGRIAS101519EA 282 EUR
AGRIAS101519
Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: Llama
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR