Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)

Lieferant: Agrisera
AS101529
AGRIAS101529EA 210 EUR
AGRIAS101529
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR