Anti-IgG (H+L) Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Agrisera
AS101537
AGRIAS101537EA 225 EUR
AGRIAS101537
Anti-IgG (H+L) Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Sheep
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR