Anti-PPO Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS101585
AGRIAS101585EA 449 EUR
AGRIAS101585
Anti-PPO Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Carica papaya
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR