Anti-ADH Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS10685
AGRIAS10685EA 428 EUR
AGRIAS10685
Anti-ADH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR