Anti-Deg5 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS10703
AGRIAS10703EA 428 EUR
AGRIAS10703
Anti-Deg5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DEG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Citrullus vulgaris, Physcomitrella patens
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR