Anti-H3 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS10710
AGRIAS10710EA 329 EUR
AGRIAS10710
Anti-H3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: R. capsulatus, R. pomeroyi dss-3
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR