Anti-IgA alpha Goat Polyclonal Antibody (Biotin)

Lieferant: Agrisera
AS10745
AGRIAS10745EA 253 EUR
AGRIAS10745
Anti-IgA alpha Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR