Anti-ADH/ALDH Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS10748
AGRIAS10748EA 428 EUR
AGRIAS10748
Anti-ADH/ALDH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Cucurbita sp.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR