Anti-IgG (H+L) Goat Polyclonal Antibody
Lieferant: Agrisera
AS10780
AGRIAS10780EA 253 EUR
AGRIAS10780
Anti-IgG (H+L) Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human