Anti-IgG F(c) Goat Polyclonal Antibody
Lieferant: Agrisera
AS10792
AGRIAS10792EA 253 EUR
AGRIAS10792
Anti-IgG F(c) Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human