Anti-IgG F(c) Goat Polyclonal Antibody (Biotin)

Lieferant: Agrisera
AS10794
AGRIAS10794EA 282 EUR
AGRIAS10794
Anti-IgG F(c) Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR