Anti-IgG (H+L) Chicken Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Agrisera
AS10835
AGRIAS10835EA 225 EUR
AGRIAS10835
Anti-IgG (H+L) Chicken Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Chicken
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR