Anti-Sml1 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS10847
AGRIAS10847EA 329 EUR
AGRIAS10847
Anti-Sml1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SML1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Synechocystis sp. pcc 6803
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR