Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: Agrisera
AS10848
AGRIAS10848EA 253 EUR
AGRIAS10848
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit