Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)

Lieferant: Agrisera
AS10849
AGRIAS10849EA 225 EUR
AGRIAS10849
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR