Anti-PsaC Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS10939
AGRIAS10939EA 329 EUR
AGRIAS10939
Anti-PsaC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PSAC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, C. reinhardtii, H. vulgare, N. tabacum, Synechococcus sp. pcc7942
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR