Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 488)
Lieferant: Agrisera
AS10949
AGRIAS10949EA 225 EUR
AGRIAS10949
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Donkey
Isotype:
Reactivity: Rat