Anti-IgG (H+L) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Agrisera
AS10964
AGRIAS10964EA 253 EUR
AGRIAS10964
Anti-IgG (H+L) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse