Anti-IgG (H+L) Goat Polyclonal Antibody

Lieferant: Agrisera
AS10982
AGRIAS10982EA 207 EUR
AGRIAS10982
Anti-IgG (H+L) Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR