Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)
Lieferant: Agrisera
AS10997
AGRIAS10997EA 268 EUR
AGRIAS10997
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Goat, Human, Mouse, Rabbit, Rat