Anti-Beta-CA1/ beta-CA2 Rabbit Polyclonal Antibody
Lieferant: Agrisera
AS111737
AGRIAS111737EA 428 EUR
AGRIAS111737
Anti-Beta-CA1/ beta-CA2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: beta CA1/2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana