Anti-Acd1 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS111783
AGRIAS111783EA 329 EUR
AGRIAS111783
Anti-Acd1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADC-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR