Anti-gamma-glutamyl-cysteine Rabbit Polyclonal Antibody
Lieferant: Agrisera
AS111811
AGRIAS111811EA 492 EUR
AGRIAS111811
Anti-gamma-glutamyl-cysteine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: gamma-glutamyl-cysteine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana