Anti-BRI1 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS121859
AGRIAS121859EA 329 EUR
AGRIAS121859
Anti-BRI1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BRI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR