Anti-CrPDAT1 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS121875
AGRIAS121875EA 428 EUR
AGRIAS121875
Anti-CrPDAT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CRPDAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR