Anti-IgG (H+L) Chicken Polyclonal Antibody (DyLight® 550)

Lieferant: Agrisera
AS121932
AGRIAS121932EA 284 EUR
AGRIAS121932
Anti-IgG (H+L) Chicken Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Chicken
Isotype:
Reactivity: Llama
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR