Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 550)

Lieferant: Agrisera
AS121935
AGRIAS121935EA 225 EUR
AGRIAS121935
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Donkey
Isotype:
Reactivity: Human, Mouse, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR