Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 550)

Lieferant: Agrisera
AS121939
AGRIAS121939EA 185 EUR
AGRIAS121939
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit, Rat, Sheep
Bestellen

TECHNISCHE DATEN

Antigen-Symbol IgG (H+L)
Environmentally Preferable
Antikörper-Typ Sekundär
Klonalität Polyklonal
Konjugation DyLight® 550
Reaktivität Hamster,Rabbit,Horse,Sheep,Goat,Chicken,Human,Bovine,Guinea pig,Rat,Mouse
Wirt Goat
Isotyp
Western blot Yes
ImmunoChemie Yes
Durchflusszytometrie Yes
Immunogen Purified Mouse IgG,, whole molecule
Reinigung purified
Kreuzadsorption No
Lagertemperatur –20 °C
Transporttemperatur Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR