Anti-PIF5 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS122112
AGRIAS122112EA 329 EUR
AGRIAS122112
Anti-PIF5 Rabbit Polyclonal Antibody
Antikörper
PIF5 (Phytochrome interacting factor 5) is a transcription factor which acts negatively in the phytochrome B signaling pathway and regulates PHYB at post-transcriptional level. Alternative protein names: AtbHLH65, bHLH65, bHLH065, Basic helix-loop-helix protein 65, PHYTOCHROME INTERACTING FACTOR 3-LIKE 6, PIL6, Phytochrome interacting factor-like 6, Transcription factor EN 103.,PIF5 (Phytochrome interacting factor 5) is a transcription factor which acts negatively in the phytochrome B signaling pathway and regulates PHYB at post-transcriptional level. Alternative protein names: AtbHLH65, bHLH65, bHLH065, Basic helix-loop-helix protein 65, PHYTOCHROME INTERACTING FACTOR 3-LIKE 6, PIL6, Phytochrome interacting factor-like 6, Transcription factor EN 103.

Type: Primary
Antigen: PIF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR