Anti-AHK2 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS122113
AGRIAS122113EA 329 EUR
AGRIAS122113
Anti-AHK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AHK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR