Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 633)

Lieferant: Agrisera
AS122236
AGRIAS122236EA 225 EUR
AGRIAS122236
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 633)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 633
Epitope:
Host: Donkey
Isotype:
Reactivity: Human, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR