Anti-IgG F(c) Goat Polyclonal Antibody (DyLight® 650)

Lieferant: Agrisera
AS122315
AGRIAS122315EA 180 EUR
AGRIAS122315
Anti-IgG F(c) Goat Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Horse, Human, Mouse, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR