Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 650)

Lieferant: Agrisera
AS122335
AGRIAS122335EA 253 EUR
AGRIAS122335
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR