Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 800)

Lieferant: Agrisera
AS122436
AGRIAS122436EA 240 EUR
AGRIAS122436
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 800)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Goat, Human, Mouse, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR