Anti-CesA4 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS122582
AGRIAS122582EA 329 EUR
AGRIAS122582
Anti-CesA4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CESA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR