Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 350)

Lieferant: Agrisera
AS122588
AGRIAS122588EA 180 EUR
AGRIAS122588
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 350)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 350
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit, Rat, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR