Anti-GID2 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS132638
AGRIAS132638EA 329 EUR
AGRIAS132638
Anti-GID2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GID2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Brassica oleracea var. italica, Brassica rapa var. komatsuna, C. reinhardtii, Citrus limon, N. tabacum, Spinacia oleracea, Zea mays
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR