Anti-LHY Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS132661
AGRIAS132661EA 329 EUR
AGRIAS132661
Anti-LHY Rabbit Polyclonal Antibody
Antikörper
LHY (Late elongated hypocotyl) is a transcription factor which is involved in the circadian clock. It binds to the promoter region of APRR1/TOC1 and TCP21/CHE to repress their transcription. Alternative names: LATE ELONGATED HYPOCOTYL, LATE ELONGATED HYPOCOTYL 1, LHY, LHY1.,LHY (Late elongated hypocotyl) is a transcription factor which is involved in the circadian clock. It binds to the promoter region of APRR1/TOC1 and TCP21/CHE to repress their transcription. Alternative names: LATE ELONGATED HYPOCOTYL, LATE ELONGATED HYPOCOTYL 1, LHY, LHY1.

Type: Primary
Antigen: LHY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR