Anti-MT2a Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS132685
AGRIAS132685EA 329 EUR
AGRIAS132685
Anti-MT2a Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MT2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR