Anti-Sec8 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS132708
AGRIAS132708EA 329 EUR
AGRIAS132708
Anti-Sec8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SEC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Zea mays
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR