Anti-ASyO2 Mouse Monoclonal Antibody
Lieferant: Agrisera
AS132717B
AGRIAS132717BEA 672 EUR
AGRIAS132717B
Anti-ASyO2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ASYO2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: A. thaliana, Spinacia oleracea, Synechocystis pcc 6803