Anti-SVR4-like Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS132726
AGRIAS132726EA 329 EUR
AGRIAS132726
Anti-SVR4-like Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SVR4-like
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, H. vulgare
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR