Anti-Acrylamid Chicken Polyclonal Antibody

Lieferant: Agrisera
AS132732
AGRIAS132732EA 597 EUR
AGRIAS132732
Anti-Acrylamid Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Acrylamide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Hordeum vulgare
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR