Anti-AHB2 Rabbit Polyclonal Antibody

Lieferant: Agrisera
AS132745
AGRIAS132745EA 329 EUR
AGRIAS132745
Anti-AHB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AHB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Guinea Pig, Human, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR