Anti-IgG (H+L) Llama Polyclonal Antibody (DyLight® 488)

Lieferant: Agrisera
AS152839
AGRIAS152839EA 202 EUR
AGRIAS152839
Anti-IgG (H+L) Llama Polyclonal Antibody (DyLight® 488)
Antikörper
Llama anti-Rabbit IgG (H&L), DyLight 488 Conjugate is a secondary antibody which binds to heavy (?) chains chains on rabbit IgG light chains on all rabbit immunoglobulins.DyLight® 488 Amax = 493 nm, Emax = 518 nm. Antibodies are purified using solid phase Rabbit IgG (H&L).DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.,Llama anti-Rabbit IgG (H&L), DyLight 488 Conjugate is a secondary antibody which binds to heavy (?) chains chains on rabbit IgG light chains on all rabbit immunoglobulins.DyLight® 488 Amax = 493 nm, Emax = 518 nm. Antibodies are purified using solid phase Rabbit IgG (H&L).DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Llama
Isotype:
Reactivity: Human, Mouse, Rabbit, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR