Anti-GST Goat Polyclonal Antibody (Europium 1024)

Lieferant: Agrisera
AS152913
AGRIAS152913EA 891 EUR
AGRIAS152913
Anti-GST Goat Polyclonal Antibody (Europium 1024)
Antikörper
Goat anti-GST is a primary antibody, which binds to GST and is directly conjugated to Europium 1024. ,Goat anti-GST is a primary antibody, which binds to GST and is directly conjugated to Europium 1024.

Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Europium 1024
Epitope:
Host: Goat
Isotype:
Reactivity: A. thaliana
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR