Anti-HA Mouse Monoclonal Antibody (SureLight® R-Phycoerythrin) [clone: 16B12]
Lieferant: Agrisera
AS152918
AGRIAS152918EA 482 EUR
AGRIAS152918
Anti-HA Mouse Monoclonal Antibody (SureLight® R-Phycoerythrin) [clone: 16B12]
Antikörper
Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to SureLight® R-Phycoerythrin. This is of advantage to shorten assay time by no need to use a secondary antibody to HA. ,Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to SureLight® R-Phycoerythrin. This is of advantage to shorten assay time by no need to use a secondary antibody to HA.
Type: Primary
Antigen: HA
Clonality: Monoclonal
Clone: 16B12
Conjugation: SureLight® RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Chlamydomonas reinhardtii