Anti-HA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Agrisera
AS152924
AGRIAS152924EA 482 EUR
AGRIAS152924
Anti-HA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to HRP. This is of advantage to shorten assay time by no need to use a secondary antibody to HA. ,Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to HRP. This is of advantage to shorten assay time by no need to use a secondary antibody to HA.
Type: Primary
Antigen: HA
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen