Anti-6x His Mouse Monoclonal Antibody (DyLight® 488) [clone: AD1.1.10]

Lieferant: Agrisera
AS152928
AGRIAS152928EA 482 EUR
AGRIAS152928
Anti-6x His Mouse Monoclonal Antibody (DyLight® 488) [clone: AD1.1.10]
Antikörper
Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to DyLight® 488. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag. DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.,Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to DyLight® 488. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag. DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.

Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.10
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype:
Reactivity: A. thaliana
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR